(3-Methoxy-4-methylphenyl)acetonitril
Lieferant: Apollo Scientific
Synonyme:
3-Methoxy-4-methylphenylacetonitrile
OR350248-5G
APOSOR350248-5GEA
62.5
EUR
APOSOR350248-5G
(3-Methoxy-4-methylphenyl)acetonitril
(3-Methoxy-4-methylphenyl)acetonitril
Formel:
C₁₀H₁₁NO Molecular Weight: 161,2004 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00060309 CAS-Nummer: 64829-31-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...