2-Methyl-3-(methylthio)pyrazin

Lieferant: Apollo Scientific

OR350253-5G
APOSOR350253-5GEA 0 EUR
APOSOR350253-5G
2-Methyl-3-(methylthio)pyrazin
2-Methyl-3-(methylthio)pyrazine
Formel: C₆H₈N₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00014610
CAS-Nummer: 2882-20-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR