3-Methylzimtsäure

Lieferant: Apollo Scientific
Achtung

OR350254-25G OR350254-5G
APOSOR350254-25GEA 56.7 EUR
APOSOR350254-25G APOSOR350254-5G
3-Methylzimtsäure
3-Methylzimtsäure
Formel: C₁₀H₁₀O₂
Schmelzpunkt: 116…119 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016844
CAS-Nummer: 3029-79-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR