Methyl 6-methylpyrazine-2-carboxylate

Lieferant: Apollo Scientific

OR350256-1G
APOSOR350256-1GEA 126 EUR
APOSOR350256-1G
Methyl 6-methylpyrazine-2-carboxylate
Methyl 6-methylpyrazine-2-carboxylate
Formel: C₇H₈N₂O₂
Molecular Weight: 152,151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08705757
CAS-Nummer: 41110-38-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR