Methyl-1-amino-1-cyclopentancarboxylat-hydrochlorid

Lieferant: Apollo Scientific

OR350260-5G
APOSOR350260-5GEA 0 EUR
APOSOR350260-5G
Methyl-1-amino-1-cyclopentancarboxylat-hydrochlorid
Methyl-1-aminocyclopentanecarboxylate hydrochloride
Formel: C₇H₁₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03840364
CAS-Nummer: 60421-23-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR