2-Methyl-3-tetrahydrothiophenone
Lieferant: Apollo Scientific
Synonyme:
Dihydro-2-methyl-3(2H)-thiophenone, 2-Methyl-3-oxotetrahydrothiophene
OR350261-25G
APOSOR350261-25GEA
79.9
EUR
APOSOR350261-25G
2-Methyl-3-tetrahydrothiophenone
2-Methyl-3-tetrahydrothiophenone
Formel:
C₅H₈OS Molecular Weight: 116,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00078280 CAS-Nummer: 13679-85-1 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...