Methyl-1,2,4-triazol-3-carboxylat 97%

Lieferant: Apollo Scientific

Synonyme: Methyl 1H-1,2,4-triazole-3-carboxylate, Methyl 1H-1,2,4-triazole-5-carboxylate

OR350262-500G
APOSOR350262-500GEA 74.1 EUR
APOSOR350262-500G
Methyl-1,2,4-triazol-3-carboxylat 97%
Methyl-1,2,4-triazol-3-carboxylat
Formel: C₄H₅N₃O₂
Molecular Weight: 127.10 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00135989
CAS-Nummer: 4928-88-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR