3-[(4-Nitrobenzyl)oxy]-3-oxopropansäure

Lieferant: Apollo Scientific

Synonyme: Mono-4-nitrobenzyl Malonate, Malonic acid mono-4-nitrobenzyl ester, 4-Nitrobenzyl Hydrogen Malonate

OR350266-100G
APOSOR350266-100GEA 63.8 EUR
APOSOR350266-100G
3-[(4-Nitrobenzyl)oxy]-3-oxopropansäure
3-((4-Nitrobenzyl)oxy)-3-oxopropanoic acid
Formel: C₁₀H₉NO₆
Molecular Weight: 239,1816 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00191556
CAS-Nummer: 77359-11-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR