Methyl-4-amino-2-chlorbenzoat

Lieferant: Apollo Scientific
Achtung

OR350268-100G
APOSOR350268-100GEA 253 EUR
APOSOR350268-100G
Methyl-4-amino-2-chlorbenzoat
Methyl-4-amino-2-chlorbenzoat
Formel: C₈H₈ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03840484
CAS-Nummer: 46004-37-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR