2-(3-Methoxybenzyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Lieferant: Apollo Scientific

Synonyme: (3-Methoxybenzyl)boronic Acid Pinacol Ester

OR350269-250MG
APOSOR350269-250MGEA 134 EUR
APOSOR350269-250MG
2-(3-Methoxybenzyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
2-(3-Methoxybenzyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formel: C₁₄H₂₁BO₃
Molecular Weight: 248,126 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10698524
CAS-Nummer: 797762-23-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR