p-Tolylacetonitril

Lieferant: Apollo Scientific

Synonyme: (4-Methylphenyl)acetonitril, 4-Methylbenzylcyanid

OR350270-100G
APOSOR350270-100GEA 77 EUR
APOSOR350270-100G
p-Tolylacetonitril
p-Tolylacetonitril
Formel: C₉H₉N
Molecular Weight: 131,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00001922
CAS-Nummer: 2947-61-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR