2-(Methylthio)phenol 98%

Lieferant: Apollo Scientific

Synonyme: 2-Hydroxythioanisole

OR350271-25G
APOSOR350271-25GEA 120 EUR
APOSOR350271-25G
2-(Methylthio)phenol 98%
2-(Methylthio)phenol
Formel: C₇H₈OS
Molecular Weight: 140.21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002211
CAS-Nummer: 1073-29-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR