5-Methoxynicotinonitrile 98%

Lieferant: Apollo Scientific

OR350274-5G
APOSOR350274-5GEA 230 EUR
APOSOR350274-5G
5-Methoxynicotinonitrile 98%
5-Methoxynicotinonitrile
Formel: C₇H₆N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD09909444
CAS-Nummer: 298204-74-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR