6-Methyl-5-nitro-2,4(1H,3H)-pyrimidindion

Lieferant: Apollo Scientific

Synonyme: 6-Methyl-5-nitrouracil, 2,4-Dihydroxy-6-methyl-5-nitropyrimidine

OR350278-5G
APOSOR350278-5GEA 58 EUR
APOSOR350278-5G
6-Methyl-5-nitro-2,4(1H,3H)-pyrimidindion
5-Nitro-6-methyluracil
Formel: C5H5N3O4
Molecular Weight: 171,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00047362
CAS-Nummer: 16632-21-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR