Methyl-3-(2-bromphenyl)propanoat
Lieferant: Apollo Scientific
Synonyme:
3-(2-Bromophenyl)propionic Acid Methyl Ester, Methyl 3-(2-bromophenyl)propanoate
OR350280-25G
APOSOR350280-25GEA
115
EUR
APOSOR350280-25G
Methyl-3-(2-bromphenyl)propanoat
Methyl-3-(2-bromphenyl)propanoat
Formel:
C₁₀H₁₁BrO₂ Molecular Weight: 243,10 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09953149 CAS-Nummer: 66191-86-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...