Morpholine-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Morpholinecarboxylic Acid

OR350283-1G
APOSOR350283-1GEA 197 EUR
APOSOR350283-1G
Morpholine-2-carboxylic acid
Morpholine-2-carboxylic acid
Formel: C₅H₉NO₃
Molecular Weight: 131,1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03788074
CAS-Nummer: 300582-83-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR