Benzyloxycarbonyl-L-Methionin
Lieferant: Apollo Scientific
Synonyme:
N-Carbobenzyloxy-L-methionine, Z-Met-OH, N-Cbz-L-methionine, Cbz-Met-OH
OR350299-100G
APOSOR350299-100GEA
115
EUR
APOSOR350299-100G
Benzyloxycarbonyl-L-Methionin
Benzyloxycarbonyl-L-Methionin
Formel:
C₁₃H₁₇NO₄S Molecular Weight: 283,34 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00065124 CAS-Nummer: 1152-62-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...