3-Brom-5-methoxyphenylboronsäure 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Bromo-5-methoxyphenyl)boronic acid

OR3503-5G
APOSOR3503-5GEA 115 EUR
APOSOR3503-5G
3-Brom-5-methoxyphenylboronsäure 96%
3-Brom-5-methoxyphenylboronsäure
Formel: C₇H₈BBrO₃
Molecular Weight: 230.8516 g/mol
Schmelzpunkt: 224…226 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07369759
CAS-Nummer: 849062-12-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR