Carbobenzyloxy-beta-Alanin

Lieferant: Apollo Scientific

Synonyme: Carbobenzyloxy-beta-alanine

OR350305-100G
APOSOR350305-100GEA 59.6 EUR
APOSOR350305-100G
Carbobenzyloxy-beta-Alanin
Carbobenzyloxy-beta-Alanin
Formel: C₁₁H₁₃NO₄
Molecular Weight: 223,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00037292
CAS-Nummer: 2304-94-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR