N-(Tert-Butoxycarbonyl)-1 H-Pyrazol-1-Carboxamidin

Lieferant: Apollo Scientific

Synonyme: 1-(N-Boc-amidino)pyrazole, N-Boc-1H-pyrazole-1-carboximidamide, 1-[N-(tert-Butoxycarbonyl)carbamimidoyl]pyrazole, N-Boc-1H-pyrazole-1-carboxamidine, N-(tert-Butoxycarbonyl)-1H-pyrazole-1-carboximidamide, 1-[N-(tert-Butoxycarbonyl)amidino]pyrazole, N-Boc-1-Guanylpyrazole

OR350307-25G
APOSOR350307-25GEA 65.4 EUR
APOSOR350307-25G
N-(Tert-Butoxycarbonyl)-1 H-Pyrazol-1-Carboxamidin
N-(Tert-Butoxycarbonyl)-1 H-Pyrazol-1-Carboxamidin
Formel: C₉H₁₄N₄O₂
Molecular Weight: 210.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00216663
CAS-Nummer: 152120-61-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR