N-Chlorphthalimid

Lieferant: Apollo Scientific

OR350308-100G
APOSOR350308-100GEA 87 EUR
APOSOR350308-100G
N-Chlorphthalimid
N-Chlorphthalimid
Formel: C₈H₄ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00023027
CAS-Nummer: 3481-09-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR