3-Piperidinmethanol
Lieferant: Apollo Scientific
Synonyme:
3-(Hydroxymethyl)piperidine
OR350315-100G
APOSOR350315-100GEA
89.9
EUR
APOSOR350315-100G
3-Piperidinmethanol
3-Piperidinmethanol
Formel:
C₆H₁₃NO Molecular Weight: 115,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005997 CAS-Nummer: 4606-65-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...