1,3-Phenylendiessigsäure
Lieferant: Apollo Scientific
OR350319-25G
APOSOR350319-25GEA
149
EUR
APOSOR350319-25G
1,3-Phenylendiessigsäure
1,3-Phenylendiessigsäure
Formel:
C₁₀H₁₀O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004341 CAS-Nummer: 19806-17-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...