8-Chinolinsulfonsäure

Lieferant: Apollo Scientific

Synonyme: 8-Quinolinesulphonic acid

OR350330-25G
APOSOR350330-25GEA 0 EUR
APOSOR350330-25G
8-Chinolinsulfonsäure
8-Chinolinsulfonsäure
Formel: C₉H₇NO₃S
Molecular Weight: 209,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00024037
CAS-Nummer: 85-48-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR