2,6-Dibromoanisole

Lieferant: Apollo Scientific

Synonyme: 1,3-Dibromo-2-methoxybenzene

OR350334-1G
APOSOR350334-1GEA 0 EUR
APOSOR350334-1G
2,6-Dibromoanisole
2,6-Dibromoanisole
Formel: C₇H₆Br₂O
Molecular Weight: 265,93 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00093270
CAS-Nummer: 38603-09-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR