(R)-4-Chloromethyl-2,2-dimethyl-1,3-dioxolane
Lieferant: Apollo Scientific
Synonyme:
(R)-4-(Chloromethyl)-2,2-dimethyl-1,3-dioxolane, (R)-(+)-4-(Chloromethyl)-2,2-dimethyl-1,3-dioxolane
OR350338-25G
APOSOR350338-25GEA
58
EUR
APOSOR350338-25G
(R)-4-Chloromethyl-2,2-dimethyl-1,3-dioxolane
(R)-4-Chloromethyl-2,2-dimethyl-1,3-dioxolane
Formel:
C₆H₁₁ClO₂ Molecular Weight: 150.60 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00273371 CAS-Nummer: 57044-24-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...