(4R,6S)-6-Methyl-7,7-dioxo-5,6-dihydro-4H-thieno[2,3-b]thiopyran-4-ol

Lieferant: Apollo Scientific

Synonyme: (4R,6S)-6-Methyl-5,6-dihydro-4H-thieno[2,3-b]thiopyran-4-ol-7,7-dioxide

OR350339-5G
APOSOR350339-5GEA 75.4 EUR
APOSOR350339-5G
(4R,6S)-6-Methyl-7,7-dioxo-5,6-dihydro-4H-thieno[2,3-b]thiopyran-4-ol
(4R,6S)-6-Methyl-7,7-dioxo-5,6-dihydro-4H-thieno[2,3-b]thiopyran-4-ol
Formel: C₈H₁₀O₃S₂
Molecular Weight: 218,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11035925
CAS-Nummer: 147128-77-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR