2-sec-Butyl-3-methoxypyrazin

Lieferant: Apollo Scientific

Synonyme: 2-(sec-Butyl)-3-methoxypyrazine

OR350347-25G
APOSOR350347-25GEA 159 EUR
APOSOR350347-25G
2-sec-Butyl-3-methoxypyrazin
2-sec-Butyl-3-methoxypyrazin
Formel: C₉H₁₄N₂O
Molecular Weight: 166,2203 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006136
CAS-Nummer: 24168-70-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR