(1S,3aR,6aS)-tert-Butyl octahydrocyclopenta[c]pyrrole-1-carboxylate oxalate

Lieferant: Apollo Scientific

OR350352-1G
APOSOR350352-1GEA 122 EUR
APOSOR350352-1G
(1S,3aR,6aS)-tert-Butyl octahydrocyclopenta[c]pyrrole-1-carboxylate oxalate
(1S,3aR,6aS)-tert-Butyl octahydrocyclopenta[c]pyrrole-1-carboxylate oxalate
Formel: C₁₄H₂₃NO₆
Molecular Weight: 301,3400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22124391
CAS-Nummer: 907606-68-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR