(S)-2-(2-Hydroxyethyl)piperidine hydrochloride

Lieferant: Apollo Scientific

OR350353-250MG
APOSOR350353-250MGEA 26.1 EUR
APOSOR350353-250MG
(S)-2-(2-Hydroxyethyl)piperidine hydrochloride
(S)-2-(2-Hydroxyethyl)piperidine hydrochloride
Formel: C₇H₁₆ClNO
Molecular Weight: 165,6610 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09265049
CAS-Nummer: 786684-21-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR