(4S)-4-(4-Aminobenzyl)-1,3-oxazolidin-2-on
Lieferant: Apollo Scientific
Synonyme:
(4S)-4-[(4-Aminophenyl)methyl]-2-oxazolidinone, (S)-4-(4-Aminobenzyl)-2(1H)-oxazolidinone
OR350358-25G
APOSOR350358-25GEA
56.7
EUR
APOSOR350358-25G
(4S)-4-(4-Aminobenzyl)-1,3-oxazolidin-2-on
(S)-4-(4-Aminobenzyl)oxazolidin-2-one
Formel:
C₁₀H₁₂N₂O₂ Molecular Weight: 192,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03411476 CAS-Nummer: 152305-23-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...