(S)-2-(Piperidin-2-yl)ethanol

Lieferant: Apollo Scientific

OR350359-250MG
APOSOR350359-250MGEA 0 EUR
APOSOR350359-250MG
(S)-2-(Piperidin-2-yl)ethanol
(S)-2-(Piperidin-2-yl)ethanol
Formel: C₇H₁₅NO
Lagertemperatur: Raumtemperatur
MDL: MFCD08461072
CAS-Nummer: 103639-57-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR