(S)-N6-Propyl-4,5,6,7-tetrahydrobenzothiazole-2,6-diamine 98%
Lieferant: Apollo Scientific
Synonyme:
(S)-N6-Propyl-4,5,6,7-tetrahydrobenzo[d]thiazole-2,6-diamine, Pramipexole
OR350364-1G
APOSOR350364-1GEA
89.9
EUR
APOSOR350364-1G
(S)-N6-Propyl-4,5,6,7-tetrahydrobenzothiazole-2,6-diamine 98%
(S)-N6-Propyl-4,5,6,7-tetrahydrobenzothiazole-2,6-diamine
Formel:
C₁₀H₁₇N₃S Molecular Weight: 211,327 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00869076 CAS-Nummer: 104632-26-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...