(S)-β-(4-Chlorphenyl)alaninol

Lieferant: Apollo Scientific

Synonyme: (S)-3-Amino-3-(4-chlorophenyl)-1-propanol, (S)-3-Amino-3-(4-chlorophenyl)propan-1-ol

OR350365-1G
APOSOR350365-1GEA 119 EUR
APOSOR350365-1G
(S)-β-(4-Chlorphenyl)alaninol
(S)-β-(4-Chlorphenyl)alaninol
Formel: C₉H₁₂ClNO
Molecular Weight: 185,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07784069
CAS-Nummer: 886061-26-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR