1,1,1-Tris(4-hydroxyphenyl)ethane

Lieferant: Apollo Scientific

OR350366-100G
APOSOR350366-100GEA 0 EUR
APOSOR350366-100G
1,1,1-Tris(4-hydroxyphenyl)ethane
1,1,1-Tris(4-hydroxyphenyl)ethane
Formel: C₂₀H₁₈O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00012180
CAS-Nummer: 27955-94-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR