trans-4-(Aminomethyl)cyclohexancarbonsäure 98+%

Lieferant: Apollo Scientific
Achtung

Synonyme: HAKU, AMCA, TAMCHA, trans-4-(aminomethyl)cyclohexane-1-carboxylic acid, Tranexamic acid, AMCHA

OR350367-100G OR350367-1KG OR350367-500G
APOSOR350367-100GEA 59.6 EUR
APOSOR350367-100G APOSOR350367-1KG APOSOR350367-500G
trans-4-(Aminomethyl)cyclohexancarbonsäure 98+%
trans-4-(Aminomethyl)cyclohexanecarboxylic acid
Formel: C₈H₁₅NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00001466
CAS-Nummer: 1197-18-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR