2-(p-Tolyl)pyridin
Lieferant: Apollo Scientific
OR350374-100G
APOSOR350374-100GEA
207
EUR
APOSOR350374-100G
2-(p-Tolyl)pyridin
2-(p-Tolyl)pyridin
Formel:
C₁₂H₁₁N Molecular Weight: 169,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006281 CAS-Nummer: 4467-06-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...