1-Boc-3-indolboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: (1-(tert-Butoxycarbonyl)-1H-indol-3-yl)boronic acid

OR350387-5G
APOSOR350387-5GEA 84.1 EUR
APOSOR350387-5G
1-Boc-3-indolboronsäure
1-Boc-3-indolboronsäure
Formel: C₁₃H₁₆BNO₄
Molecular Weight: 261.0814 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD06656257
CAS-Nummer: 181365-26-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR