5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(2-((trimethylsilyl)oxy)propan-2-yl)pyridine

Lieferant: Apollo Scientific

OR350390-1G
APOSOR350390-1GEA 77 EUR
APOSOR350390-1G
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(2-((trimethylsilyl)oxy)propan-2-yl)pyridine
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(2-((trimethylsilyl)oxy)propan-2-yl)pyridine
Formel: C₁₇H₃₀BNO₃Si
Molecular Weight: 335,321 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12198135
CAS-Nummer: 1228014-10-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR