5,6,7,8-Tetrahydro-1,6-naphthyridine hydrochloride

Lieferant: Apollo Scientific

Synonyme: 5,6,7,8-Tetrahydro-1,6-naphthyridine hcl

OR350392-100MG
APOSOR350392-100MGEA 92.8 EUR
APOSOR350392-100MG
5,6,7,8-Tetrahydro-1,6-naphthyridine hydrochloride
5,6,7,8-Tetrahydro-1,6-naphthyridine hydrochloride
Formel: C₈H₁₁ClN₂
Molecular Weight: 170,64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11846187
CAS-Nummer: 1187830-51-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR