4-(Trimethylsilyl)phenylboronsäure 95%

Lieferant: Apollo Scientific

Synonyme: (4-(Trimethylsilyl)phenyl)boronic acid

OR350393-10G
APOSOR350393-10GEA 105 EUR
APOSOR350393-10G
4-(Trimethylsilyl)phenylboronsäure 95%
4-(Trimethylsilyl)phenylboronsäure
Formel: C₉H₁₅BO₂Si
Molecular Weight: 194,1100 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00093348
CAS-Nummer: 17865-11-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR