Tris(4-nitrophenyl)amine
Lieferant: Apollo Scientific
OR350395-100G
APOSOR350395-100GEA
163
EUR
APOSOR350395-100G
Tris(4-nitrophenyl)amine
Tris(4-nitrophenyl)amine
Formel:
C₁₈H₁₂N₄O₆ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00024636 CAS-Nummer: 20440-93-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...