8-Acetyl-6-(benzyloxy)-2H-benzo[b][1,4]oxazin-3(4H)-one 97%
Lieferant: Apollo Scientific
OR350399-1G
APOSOR350399-1GEA
168
EUR
APOSOR350399-1G
8-Acetyl-6-(benzyloxy)-2H-benzo[b][1,4]oxazin-3(4H)-one 97%
8-Acetyl-6-(benzyloxy)-2H-benzo[b][1,4]oxazin-3(4H)-one
Formel:
C₁₇H₁₅NO₄ Molecular Weight: 297,305 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD24038851 CAS-Nummer: 869478-09-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...