Benzo[b]thiophene-6-carbonitrile 95+%

Lieferant: Apollo Scientific

OR350409-250MG
APOSOR350409-250MGEA 126 EUR
APOSOR350409-250MG
Benzo[b]thiophene-6-carbonitrile 95+%
Benzo[b]thiophene-6-carbonitrile
Formel: C₉H₅NS
Molecular Weight: 159,208 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16877454
CAS-Nummer: 154650-81-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR