3-Boc-9-oxo-3-azaspiro[5.5]undecane 97%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 9-oxo-3-azaspiro[5.5]undecane-3-carboxylate

OR350440-250MG
APOSOR350440-250MGEA 121 EUR
APOSOR350440-250MG
3-Boc-9-oxo-3-azaspiro[5.5]undecane 97%
3-Boc-9-oxo-3-azaspiro[5.5]undecane
Formel: C₁₅H₂₅NO₃
Molecular Weight: 267,37 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12198558
CAS-Nummer: 873924-08-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR