6-Chloro-2-(pyridin-2-yl)pyrimidin-4-amine 97%

Lieferant: Apollo Scientific

OR350451-250MG
APOSOR350451-250MGEA 81.2 EUR
APOSOR350451-250MG
6-Chloro-2-(pyridin-2-yl)pyrimidin-4-amine 97%
6-Chloro-2-(pyridin-2-yl)pyrimidin-4-amine
Formel: C₉H₇ClN₄
Molecular Weight: 206,632 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14706108
CAS-Nummer: 1014720-73-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR