(5-Chloro-2-methyl-1H-indol-3-yl)acetic acid 98%

Lieferant: Apollo Scientific

Synonyme: 2-(5-Chloro-2-methyl-1H-indol-3-yl)acetic acid

OR350462-1G
APOSOR350462-1GEA 122 EUR
APOSOR350462-1G
(5-Chloro-2-methyl-1H-indol-3-yl)acetic acid 98%
(5-Chloro-2-methyl-1H-indol-3-yl)acetic acid
Formel: C₁₁H₁₀ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00152035
CAS-Nummer: 19017-52-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR