Ethyl 6-amino-5-chloropyrimidine-4-carboxylate ≥98%
Lieferant: Apollo Scientific
OR350472-250MG
APOSOR350472-250MGEA
183
EUR
APOSOR350472-250MG
Ethyl 6-amino-5-chloropyrimidine-4-carboxylate ≥98%
Ethyl 6-amino-5-chloropyrimidine-4-carboxylate
Formel:
C₇H₈ClN₃O₂ Molecular Weight: 201,61 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22576178 CAS-Nummer: 1097250-86-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...