2-(1H-Indol-3-yl)-2-methylpropanoic acid 98%

Lieferant: Apollo Scientific

OR350487-250MG
APOSOR350487-250MGEA 221 EUR
APOSOR350487-250MG
2-(1H-Indol-3-yl)-2-methylpropanoic acid 98%
2-(1H-Indol-3-yl)-2-methylpropanoic acid
Formel: C₁₂H₁₃NO₂
Molecular Weight: 203,237 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17011803
CAS-Nummer: 2770-92-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR