Methyl 5-chloro-6-methylpyrazine-2-carboxylate

Lieferant: Apollo Scientific
Achtung

OR350502-1G
APOSOR350502-1GEA 108 EUR
APOSOR350502-1G
Methyl 5-chloro-6-methylpyrazine-2-carboxylate
Methyl 5-chloro-6-methylpyrazine-2-carboxylate
Formel: C₇H₇ClN₂O₂
Molecular Weight: 186,596 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18633146
CAS-Nummer: 77168-85-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR